accessibility menu, dialog, popup

UserName

CAS RN 114524-80-0

CH3HNOOCH3H2N

CAS RN 114524-80-0

IUPAC Name: methyl 2-amino-3-(1H-indol-3-yl)-2-methylpropanoate
Synonyms: methyl 2-amino-3-(1H-indol-3-yl)-2-methylpropanoate
Molecular Weight (g/mol): 232.28
Molecular Formula: C13H16N2O2
InChi Key: RCUNGDZWHFRBBP-UHFFFAOYNA-N
SMILES: COC(=O)C(C)(N)CC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results