missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-amino-3-(1H-indol-3-yl)-2-methylpropanoate, 97%, Maybridge™

Product Code. 10564885
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10564885

Brand: Thermo Scientific Maybridge RJC01802DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 114524-80-0
Molecular Formula C13H16N2O2
Molecular Weight (g/mol) 232.283
MDL Number MFCD00152138
InChI Key RCUNGDZWHFRBBP-UHFFFAOYSA-N
Synonym methyl 2-amino-3-1h-indol-3-yl-2-methylpropanoate, alpha-methyl-dl-tryptophan methyl ester, tryptophan,, a-methyl-, methyl ester, alpha-methyl-d,l-tryptophan methyl ester, methyl 2-azanyl-3-1h-indol-3-yl-2-methyl-propanoate, maybridge1_006684, acmc-20ekci, pubchem14800, h-me-dl-trp-ome, h-2-me-dl-try-ome
PubChem CID 2795140
IUPAC Name methyl 2-amino-3-(1H-indol-3-yl)-2-methylpropanoate
SMILES CC(CC1=CNC2=CC=CC=C21)(C(=O)OC)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 232.28
Percent Purity 97%
Chemical Name or Material Methyle2-amino-3-(1H-indol-3-yl)-2-methylpropanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.