accessibility menu, dialog, popup

UserName

CAS RN 115-69-5

CH3H2NOHHO

CAS RN 115-69-5

IUPAC Name: 2-amino-2-methylpropane-1,3-diol
Synonyms: AMPD isobutandiol-2-amine aminoglycol pentaerythritol dichlorohydrin aminomethyl propanediol gentimon
Molecular Weight (g/mol): 105.14
Molecular Formula: C4H11NO2
InChi Key: UXFQFBNBSPQBJW-UHFFFAOYSA-N
SMILES: CC(N)(CO)CO
Molecular Weight (g/mol) 
  • (7)
  • (3)
Physical Form 
  • (3)
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (7)
  • (7)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)