CAS RN 115199-26-3
CAS RN 115199-26-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2-(2-(Bromomethyl)phenyl)-2-(methoxyimino)acetate, TRC
CAS: 115199-26-3 Molecular Formula: C11H12BrNO3 Molecular Weight (g/mol): 286.12 Synonym: 2-(Bromomethyl)phenylglyoxylic Acid Methyl Ester O-Methyloxime,2-Methoxyimino-2-[2-(bromomethyl)phenyl]acetic Acid Methyl Ester,2-[2-(Bromomethyl)phenyl]-2-(methoxyimino)acetic Acid Methyl Ester,Methyl 2-(2-Bromomethylphenyl)-2-methoximinoacetate,Methyl 2-(2-Bromomethylphenyl)glyoxylate O-Methyloxime,Methyl 2-bromomethyl-alpha-methoxyiminophenylacetate IUPAC Name: methyl (2Z)-2-[2-(bromomethyl)phenyl]-2-methoxyiminoacetate SMILES: CO\N=C(/C(=O)OC)\c1ccccc1CBr