accessibility menu, dialog, popup

UserName

CAS RN 116153-81-2

ONNHOOH

CAS RN 116153-81-2

IUPAC Name: 5-(furan-2-yl)-1H-pyrazole-3-carboxylic acid
Synonyms: 5-(furan-2-yl)-1H-pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 178.15
Molecular Formula: C8H6N2O3
InChi Key: GKPSFQIKCROJOB-UHFFFAOYSA-N
SMILES: OC(=O)C1=NNC(=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)