missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10421334
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10421334 5 g 5g
10095822 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10421334 Supplier Thermo Scientific Maybridge Supplier No. CC35001DE
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 116153-81-2
Molecular Formula C8H6N2O3
Molecular Weight (g/mol) 178.147
MDL Number MFCD05170057
InChI Key GKPSFQIKCROJOB-UHFFFAOYSA-N
Synonym 5-2-furyl-1h-pyrazole-3-carboxylic acid, 5-furan-2-yl-1h-pyrazole-3-carboxylic acid, 5-furan-2-yl-2h-pyrazole-3-carboxylic acid, 3-furan-2-yl-1h-pyrazole-5-carboxylic acid, 3-2-furyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl, 5-2-furyl pyrazole-3-carboxylic acid, acmc-20f1sn, 3-2-furyl pyrazole-5-carboxylic acid
PubChem CID 654018
IUPAC Name 5-(furan-2-yl)-1H-pyrazole-3-carboxylic acid
SMILES C1=COC(=C1)C2=CC(=NN2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 178.15
Percent Purity 97%
Chemical Name or Material 5-(2-Furyl)-1H-pyrazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.