CAS RN 1162-54-5
CAS RN 1162-54-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Dehydroprogesterone, TRC
CAS: 1162-54-5 Molecular Formula: C21 H28 O2 Molecular Weight (g/mol): 312.45 Synonym: Pregna-1,4-diene-3,20-dione,1,2-Dehydroprogesterone,1-Dehydroprogesterone,NSC 63538,δ1-Progesterone,Progesterone Imp. J (EP) IUPAC Name: (8S,9S,10R,13S,14S,17S)-17-acetyl-10,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one SMILES: CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C