missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dehydroprogesterone, TRC

Product Code. 30011543
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011543 10 mg 10mg
30006701 5 mg 5mg
30008530 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011543 Supplier Toronto Research Chemicals Supplier No. TRCD23033010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1162-54-5
Molecular Formula C21 H28 O2
Molecular Weight (g/mol) 312.45
Synonym Pregna-1,4-diene-3,20-dione, 1,2-Dehydroprogesterone, 1-Dehydroprogesterone, NSC 63538, δ1-Progesterone, Progesterone Imp. J (EP)
IUPAC Name (8S,9S,10R,13S,14S,17S)-17-acetyl-10,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H28O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h8,10,12,16-19H,4-7,9,11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1
Formula Weight 312.2089
Chemical Name or Material 1,2-Dehydroprogesterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.