CAS RN 1164462-05-8
CAS RN 1164462-05-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TG4-155, MedChemExpress
MedChemExpress TG4-155 is a potent, brain-permeant and selective EP2 receptor antagonist with a Ki of 9.9 nM. TG4-155 shows low nanomolar antagonist activity against only EP2 and DP1. TG4-155 has an EP2 Schild KB of 2.4 nM and displays 550-4750-fold selectivity for EP2 over EP1, EP3, EP4 and IP, but only 14-fold selectivity against the DP1 receptor.