accessibility menu, dialog, popup

UserName

CAS RN 116668-47-4

H2NOO

CAS RN 116668-47-4

IUPAC Name: 6-aminonaphthalene-2-carboxylate
Synonyms: 6-aminonaphthalene-2-carboxylate
Molecular Weight (g/mol): 186.19
Molecular Formula: C11H8NO2
InChi Key: NZTPZUIIYNYZKT-UHFFFAOYSA-M
SMILES: NC1=CC=C2C=C(C=CC2=C1)C([O-])=O

 No Refinements