accessibility menu, dialog, popup

UserName

CAS RN 1167418-13-4

BOOH3CCH3H3CH3CNN

CAS RN 1167418-13-4

IUPAC Name: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline
Synonyms: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline
Molecular Weight (g/mol): 256.11
Molecular Formula: C14H17BN2O2
InChi Key: ZYWICCYXTGRUNM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C2N=CC=NC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results