missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline-6-boronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11976751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11976751 1 g 1g
11986751 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976751 Supplier Thermo Scientific Acros Supplier No. 445520010
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1167418-13-4
Molecular Formula C14H17BN2O2
Molecular Weight (g/mol) 256.11
MDL Number MFCD11054040
InChI Key ZYWICCYXTGRUNM-UHFFFAOYSA-N
Synonym 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxaline-6-boronic acid, pinacol ester, 6-quinoxaline boronic acid pinacol ester, quinoxalin-6-ylboronic acid pinacol ester, quinoxaline, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 44118298
IUPAC Name 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline
SMILES CC1(C)OB(OC1(C)C)C1=CC2=NC=CN=C2C=C1

Specifications

Boiling Point 130°C to 145°C (0.6 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 256.11
Percent Purity 97%
Chemical Name or Material Quinoxaline-6-boronic acid pinacol ester
compliance-icons
Product Identifier
  • Quinoxaline-6-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.