accessibility menu, dialog, popup

UserName

CAS RN 117-08-8

ClClClOOOCl

CAS RN 117-08-8

IUPAC Name: tetrachloro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: tetrachlorophthalic anhydride etrachlorophthalic acid anhydride tetrathal
Molecular Weight (g/mol): 285.89
Molecular Formula: C8Cl4O3
InChi Key: AUHHYELHRWCWEZ-UHFFFAOYSA-N
SMILES: ClC1=C2C(=O)OC(=O)C2=C(Cl)C(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)