missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorophthalic anhydride, 98%

Product Code. 10408510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10408510 250 g Plastic bottle 250g
10225680 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10408510 Supplier Thermo Scientific Acros Supplier No. 157792500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 117-08-8 | C8Cl4O3 | 285.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-08-8 , 118-74-1
Molecular Formula C8Cl4O3
Molecular Weight (g/mol) 285.89
MDL Number MFCD00005920
InChI Key AUHHYELHRWCWEZ-UHFFFAOYSA-N
Synonym tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202
PubChem CID 8326
ChEBI CHEBI:59097
IUPAC Name 4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O

Specifications

Melting Point 254.0°C to 258.0°C
Color White
Boiling Point 349.0°C to 354.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 250 g
Beilstein 17, 484
Fieser 15,300
Solubility Information Solubility in water: 0.8 mg/l (21°C). Other solubilities: hydrolysis to acid in water
Formula Weight 285.89
Percent Purity 98%
Insoluble Matter 0.5% max. (10 g in 100 ml 1 M NaOH)
Physical Form Flakes
Chemical Name or Material Tetrachlorophthalic anhydride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.