accessibility menu, dialog, popup

UserName

CAS RN 117-61-3

IUPAC Name: 4,4'-diamino-[1,1'-biphenyl]-2,2'-disulfonic acid
Synonyms: 4,4'-diamino-[1,1'-biphenyl]-2,2'-disulfonic acid
Molecular Weight (g/mol): 344.36
Molecular Formula: C12H12N2O6S2
InChi Key: MBJAPGAZEWPEFB-UHFFFAOYSA-N
SMILES: NC1=CC=C(C2=CC=C(N)C=C2S(O)(=O)=O)C(=C1)S(O)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results