accessibility menu, dialog, popup

UserName

H3N

CAS RN 118-31-0

IUPAC Name: (naphthalen-1-yl)methanaminium
Synonyms: naphthalen-1-ylmethanaminium
Molecular Weight (g/mol): 158.22
Molecular Formula: C11H12N
InChi Key: NVSYANRBXPURRQ-UHFFFAOYSA-O
SMILES: [NH3+]CC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)