missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenemethylamine, 97%

Product Code. 10722243 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722243 5 g Glass bottle 5g
10530892 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10722243 Supplier Thermo Scientific Acros Supplier No. 155580050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-31-0 | C11H12N | 158.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-31-0
Molecular Formula C11H12N
Molecular Weight (g/mol) 158.22
MDL Number MFCD00004048
InChI Key NVSYANRBXPURRQ-UHFFFAOYSA-O
Synonym 1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride
PubChem CID 8355
IUPAC Name naphthalen-1-ylmethanamine
SMILES [NH3+]CC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.0730g/mL
Boiling Point 290°C to 293°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH2NH2
Refractive Index 1.643
Quantity 5 g
Beilstein 12, 1316
Specific Gravity 1.073
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol,ether,carbon disulfide,sulfu,ric acid
Formula Weight 157.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Naphthalenemethylamine
Show More Show Less
compliance-icons
Product Identifier
  • 1-Naphthalenemethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.