accessibility menu, dialog, popup

UserName

CH3H3CS

CAS RN 118-72-9

IUPAC Name: (2,6-dimethylphenyl)sulfanide
Synonyms: (2,6-dimethylphenyl)sulfanide
Molecular Weight (g/mol): 137.22
Molecular Formula: C8H9S
InChi Key: QCLJODDRBGKIRW-UHFFFAOYSA-M
SMILES: CC1=CC=CC(C)=C1[S-]
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results