CAS RN 1184919-23-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deschloro Dasatinib, TRC
CAS: 1184919-23-0 Molecular Formula: C22 H27 N7 O2 S Molecular Weight (g/mol): 453.56 Synonym: 5-Thiazolecarboxamide, 2-[[6-[4-(2-hydroxyethyl)-1-piperazinyl]-2-methyl-4-pyrimidinyl]amino]-N-(2-methylphenyl)-,Deschlorodasatinib IUPAC Name: 2-[[6-[4-(2-hydroxyethyl)piperazin-1-yl]-2-methylpyrimidin-4-yl]amino]-N-(2-methylphenyl)-1,3-thiazole-5-carboxamide SMILES: Cc1ccccc1NC(=O)c2cnc(Nc3cc(nc(C)n3)N4CCN(CCO)CC4)s2