accessibility menu, dialog, popup

UserName

CH3CH3H3CSiCH3CH3ClNHN

CAS RN 1185092-02-7

IUPAC Name: 1H-imidazole; tert-butyl(chloro)dimethylsilane
Synonyms: imidazole; t-butyldimethylchlorosilane
Molecular Weight (g/mol): 218.8
Molecular Formula: C9H19ClN2Si
InChi Key: IFLPAOFWVBWJPG-UHFFFAOYSA-N
SMILES: N1C=CN=C1.CC(C)(C)[Si](C)(C)Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results