missing translation for 'onlineSavingsMsg'
Learn More

BDCS, silylation reagent, AcroSeal™, Thermo Scientific™

Product Code. 10410481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10410481 100 mL AcroSeal™ Glass bottle 100mL
10052450 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10410481 Supplier Thermo Scientific Acros Supplier No. 200941000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185092-02-7
Molecular Formula C6H15ClSi·C3H4N2
Molecular Weight (g/mol) 218.8
MDL Number MFCD00000501
InChI Key IFLPAOFWVBWJPG-UHFFFAOYSA-N
Synonym bdcs silylation reagent, bdcs, silylation reagent, acroseal, him; t-butyldimethylchlorosilane, tert-butyldimethylchlorosilane imidazole, t-butyl-dimethylsilyl chloride-imidazole, imidazole; t-butyldimethylchlorosilane, tert-butyl-chloro-dimethylsilane; 1h-imidazole
PubChem CID 2724131
IUPAC Name tert-butyl-chloro-dimethylsilane;1H-imidazole
SMILES CC(C)(C)[Si](C)(C)Cl.C1=CN=CN1

Specifications

Color Yellow
Density 0.9590g/mL
Flash Point 58°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4400 to 1.4420
Quantity 100 mL
Fieser 07,184; 01,489; 03,157; 08,255
Specific Gravity 0.959
Formula Weight 218.8
Physical Form Solution
Chemical Name or Material BDCS
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • BDCS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.