accessibility menu, dialog, popup

UserName

NH2OHNOHOOHO(Z)

CAS RN 118909-22-1

IUPAC Name: (2Z)-but-2-enedioic acid; 9-amino-1,2,3,4-tetrahydroacridin-4-ol
Synonyms: 9-amino-1,2,3,4-tetrahydroacridin-4-ol; maleic acid
Molecular Weight (g/mol): 330.34
Molecular Formula: C17H18N2O5
InChi Key: JUEUNEKLNSILBZ-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C(O)=O.NC1=C2C=CC=CC2=NC2=C1CCCC2O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results