missing translation for 'onlineSavingsMsg'
Learn More

Hydroxytacrine maleate salt, Thermo Scientific Chemicals

Product Code. 15483409
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483409 250 mg 250mg
15473409 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483409 Supplier Thermo Scientific Alfa Aesar Supplier No. J60680.MD
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Cholinesterase inhibitor. Potential Alzheimer's disease therapeutic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Cholinesterase inhibitor
  • Potential Alzheimer's disease therapeutic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118909-22-1
Molecular Formula C17H18N2O5
Molecular Weight (g/mol) 330.34
MDL Number MFCD00069287
InChI Key NEEKVKZFYBQFGT-BTJKTKAUSA-N
Synonym velnacrine maleate, 9-amino-1,2,3,4-tetrahydroacridin-1-ol maleate, hydroxytacrine maleate, mentane, 1-hydroxy tacrine maleate, unii-wr9885gals, velnacrine maleate usan, wr9885gals, dsstox_cid_25158
PubChem CID 5702293
IUPAC Name 9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid
SMILES C1CC(C2=C(C3=CC=CC=C3N=C2C1)N)O.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 171°C to 173°C
Color White
Quantity 250 mg
Formula Weight 330.34
Physical Form Solid
Chemical Name or Material Hydroxytacrine maleate salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.