CAS RN 1189915-73-8
CAS RN 1189915-73-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dihydroxyphenylacetic Acid-13C,18O2, TRC
CAS: 1189915-73-8 Molecular Formula: C713CH8O218O2 Molecular Weight (g/mol): 173.14 Synonym: 3,4-Dihydroxybenzeneacetic Acid-13C,18O2,(3,4-Dihydroxyphenyl)acetic Acid-13C,18O2,Ba 2773-13C,18O2,DOPAC-13C,18O2,Dihydroxyphenylacetic Acid-13C,18O2,Dopacetic Acid-13C,18O2,Homoprotocatechuic Acid-13C,18O2; SMILES: OC1=CC(C[13C]([18OH])=[18O])=CC=C1O