missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxyphenylacetic Acid-13C,18O2, TRC

Product Code. 30136633
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136633 10 mg 10mg
30008695 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136633 Supplier Toronto Research Chemicals Supplier No. TRCD45425210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189915-73-8
Molecular Formula C713CH8O218O2
Molecular Weight (g/mol) 173.14
Synonym 3,4-Dihydroxybenzeneacetic Acid-13C,18O2, (3,4-Dihydroxyphenyl)acetic Acid-13C,18O2, Ba 2773-13C,18O2, DOPAC-13C,18O2, Dihydroxyphenylacetic Acid-13C,18O2, Dopacetic Acid-13C,18O2, Homoprotocatechuic Acid-13C,18O2;
SMILES OC1=CC(C[13C]([18OH])=[18O])=CC=C1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H8O4/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3,9-10H,4H2,(H,11,12)/i8+1,11+2,12+2
Chemical Name or Material 3,4-Dihydroxyphenylacetic Acid-13C,18O2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.