CAS RN 1189931-96-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Luminol-13C4, TRC
CAS: 1189931-96-1 Molecular Formula: C413C4H7N3O2 Molecular Weight (g/mol): 181.13 SMILES: N[13c]1[13cH][13cH][13cH]c2C(=O)NNC(=O)c12