missing translation for 'onlineSavingsMsg'
Learn More

Luminol-13C4, TRC

Product Code. 30478629
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478629 0.5 mg 0.5mg
30471064 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478629 Supplier Toronto Research Chemicals Supplier No. TRCL4744520.5MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189931-96-1
Molecular Formula C413C4H7N3O2
Molecular Weight (g/mol) 181.13
SMILES N[13c]1[13cH][13cH][13cH]c2C(=O)NNC(=O)c12

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C8H7N3O2/c9-5-3-1-2-4-6(5)8(13)11-10-7(4)12/h1-3H,9H2,(H,10,12)(H,11,13)/i1+1,2+1,3+1,5+1
Chemical Name or Material Luminol-13C4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.