accessibility menu, dialog, popup

UserName

CH3SiClClN

CAS RN 1190-16-5

IUPAC Name: 4-[dichloro(methyl)silyl]butanenitrile
Synonyms: 4-[dichloro(methyl)silyl]butanenitrile
Molecular Weight (g/mol): 182.12
Molecular Formula: C5H9Cl2NSi
InChi Key: UIFBMBZYGZSWQE-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)CCCC#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results