missing translation for 'onlineSavingsMsg'
Learn More

Cyanopropylmethyldichlorosilane, 97%, Thermo Scientific™

Product Code. 10656460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656460 5 mL Glass bottle 5mL
10139649 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656460 Supplier Thermo Scientific Acros Supplier No. 432250050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1190-16-5
Molecular Formula C5H9Cl2NSi
Molecular Weight (g/mol) 182.12
MDL Number MFCD00019889
InChI Key UIFBMBZYGZSWQE-UHFFFAOYSA-N
Synonym 3-cyanopropyl methyldichlorosilane, 3-cyanopropylmethyldichlorosilane, 4-dichloromethylsilyl-butyronitrile, 3-cyanopropyldichloromethylsilane, butanenitrile, 4-dichloromethylsilyl, unii-a09zd6eimj, 4-dichloromethylsilyl butyronitrile, methylcyanopropyldichlorosilane, 4-dichloro methyl silyl butanenitrile, dichlor-3-kyanpropyl-methylsilan
PubChem CID 14479
IUPAC Name 4-[dichloro(methyl)silyl]butanenitrile
SMILES C[Si](Cl)(Cl)CCCC#N

Specifications

Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 93.0°C to 94.0°C (8.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.15
Formula Weight 182.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyanopropylmethyldichlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyanopropylmethyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.