CAS RN 1191-67-9
CAS RN 1191-67-9
IUPAC Name:
decane-1,10-dithiol
Synonyms:
decane-1,10-dithiol
Molecular Weight (g/mol):
206.41
Molecular Formula:
C10H22S2
InChi Key:
UOQACRNTVQWTFF-UHFFFAOYSA-N
SMILES:
SCCCCCCCCCCS
1
–
1
of
1
results
1
1,10-Decanedithiol, 95%
CAS: 1191-67-9 Molecular Formula: C10H22S2 Molecular Weight (g/mol): 206.41 MDL Number: MFCD00022095 InChI Key: UOQACRNTVQWTFF-UHFFFAOYSA-N Synonym: 1,10-decanedithiol,1,10-dimercaptodecane,acmc-1bvry,1,10-didecyl mercaptan,decamethylene dimercaptan PubChem CID: 14494 IUPAC Name: decane-1,10-dithiol SMILES: SCCCCCCCCCCS