missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanedithiol, 95%

Product Code. 11347526
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11347526

Brand: Thermo Scientific Alfa Aesar L11064.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1191-67-9 | C10H22S2 | 206.41 g/mol

1,10-Decanedithiol is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Decanedithiol is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-67-9
Molecular Formula C10H22S2
Molecular Weight (g/mol) 206.41
MDL Number MFCD00022095
InChI Key UOQACRNTVQWTFF-UHFFFAOYSA-N
Synonym 1,10-decanedithiol, 1,10-dimercaptodecane, acmc-1bvry, 1,10-didecyl mercaptan, decamethylene dimercaptan
PubChem CID 14494
IUPAC Name decane-1,10-dithiol
SMILES SCCCCCCCCCCS

Specifications

Melting Point ∼17°C
Density 0.95
Boiling Point 171°C to 172°C (0.5 mmHg)
Odor Stench
Refractive Index 1.499
Quantity 5 g
UN Number UN3334
Beilstein 1738606
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 206.41
Percent Purity 95%
Chemical Name or Material 1,10-Decanedithiol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.