accessibility menu, dialog, popup

UserName

CAS RN 119229-74-2

NClFHCl

CAS RN 119229-74-2

IUPAC Name: 4-chloro-3-fluoropyridine hydrochloride
Synonyms: 4-chloro-3-fluoropyridine hydrochloride
Molecular Weight (g/mol): 167.99
Molecular Formula: C5H4Cl2FN
InChi Key: YWWXNVPPEXEZPT-UHFFFAOYSA-N
SMILES: Cl.FC1=C(Cl)C=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results