missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluoropyridine hydrochloride, 97%, Thermo Scientific™

Product Code. 11486340
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486340 25 g 25g
11466340 1 g 1g
11476340 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486340 Supplier Thermo Scientific Alfa Aesar Supplier No. H27184.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-3-fluoropyridine hydrochloride is employed for the deprotection of phenol ethers.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119229-74-2
Molecular Formula C5H4Cl2FN
Molecular Weight (g/mol) 167.992
MDL Number MFCD03453233
InChI Key YWWXNVPPEXEZPT-UHFFFAOYSA-N
Synonym 4-chloro-3-fluoropyridine hydrochloride, 4-chloro-3-fluoropyridine hcl, pyridine, 4-chloro-3-fluoro-, hydrochloride, 3-fluoro-4-chloropyridine hydrochloride, pubchem6668, acmc-20a2hg, 4-chloro-3-fluoropyridine, chloride, 4-chloro-3-fluoro pyridine hydrochloride, 4-chloranyl-3-fluoranyl-pyridine hydrochloride
PubChem CID 19360234
IUPAC Name 4-chloro-3-fluoropyridine;hydrochloride
SMILES C1=CN=CC(=C1Cl)F.Cl

Specifications

Melting Point ∼160°C (sublimation)
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 168
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-fluoropyridine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.