accessibility menu, dialog, popup

UserName

FNCO

CAS RN 1195-45-5

IUPAC Name: 1-fluoro-4-isocyanatobenzene
Synonyms: 1-fluoro-4-isocyanatobenzene
Molecular Weight (g/mol): 137.11
Molecular Formula: C7H4FNO
InChi Key: DSVGFKBFFICWLZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)