accessibility menu, dialog, popup

UserName

CH3OBOH3CH3CH3CCH3O

CAS RN 1195-66-0

IUPAC Name: 2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 158
Molecular Formula: C7H15BO3
InChi Key: JZZJAWSMSXCSIB-UHFFFAOYSA-N
SMILES: COB1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results