missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%

Product Code. 11379595
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379595 25 g 25g
11369595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379595 Supplier Thermo Scientific Alfa Aesar Supplier No. L19056.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1195-66-0 | C7H15BO3 | 158.00 g/mol

It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.

Notes
Moisture sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-66-0
Molecular Formula C7H15BO3
Molecular Weight (g/mol) 158.00
MDL Number MFCD03093086
InChI Key JZZJAWSMSXCSIB-UHFFFAOYSA-N
Synonym methoxyboronic acid pinacol ester, methoxyboronic acid, pinacol ester, 2-methoxy-4,4,5,5-tetramethyl 1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-methoxy-4,4,5,5-tetramethyl, zlchem 167, methyl pinacolyl borate, acmc-209a3a, ksc498e3h, methoxy boronic acid pinacol ester, 2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborole
PubChem CID 10975759
IUPAC Name 2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES COB1OC(C)(C)C(C)(C)O1

Specifications

Density 0.966
Boiling Point 120°C (228 mmHg)
Flash Point 46°C (114°F)
Refractive Index 1.41
Quantity 25 g
UN Number UN1993
Beilstein 2827758
Sensitivity Moisture sensitive
Formula Weight 158.01
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.