accessibility menu, dialog, popup

UserName

CAS RN 120-46-7

OO

CAS RN 120-46-7

IUPAC Name: 1,3-diphenylpropane-1,3-dione
Synonyms: dibenzoylmethane
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: NZZIMKJIVMHWJC-UHFFFAOYSA-N
SMILES: O=C(CC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)