CAS RN 12037-29-5
Praseodymium(III, IV) oxide, 99.5% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium(III, IV) oxide, 99.9% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium(III, IV) oxide, REacton™, 99.996% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium(III, IV) oxide, 99% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Praseodymium(III, IV) oxide, REacton™, 99.99% (REO)
CAS: 12037-29-5 Molecular Formula: O11Pr6 Molecular Weight (g/mol): 1021.44 MDL Number: MFCD00011178 InChI Key: HPZIIFVSYNLWNX-UHFFFAOYSA-N IUPAC Name: hexapraseodymium(3+) undecaoxidandiide SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]