missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III,IV) oxide, 99.9%, (trace metal basis)

Product Code. 10266630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
50g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10266630 50g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10266630 Supplier Thermo Scientific Acros Supplier No. 194620500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12037-29-5 | O11Pr6 | 1021.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 12037-29-5
Color Black or Brown to Green
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.90%
Molecular Formula O11Pr6
Linear Formula Pr6O11
MDL Number MFCD00011178
Merck Index 14, 7712
Solubility Information Solubility in water: insoluble
InChI Key HPZIIFVSYNLWNX-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
IUPAC Name hexapraseodymium(3+) undecaoxidandiide
Molecular Weight (g/mol) 1021.44
Formula Weight 1021.44
Percent Purity 99.9% (trace metals basis)
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Praseodymium(III, IV) oxide, 99.9%
Show More Show Less
compliance-icons
Product Identifier
  • Praseodymium(III,IV) oxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.