accessibility menu, dialog, popup

UserName

ZnAsAs

CAS RN 12044-55-2

IUPAC Name: 1λ²,2λ²-diarsenyl zinc
Synonyms: 1λ²,2λ²-diarsenyl zinc
Molecular Weight (g/mol): 215.22
Molecular Formula: As2Zn
InChi Key: KXUSEUKXCIKTBF-UHFFFAOYSA-N
SMILES: [Zn].[As]=[As]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results