missing translation for 'onlineSavingsMsg'
Learn More

Zinc arsenide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11313228
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313228 10 g 10g
11323228 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11313228 Supplier Thermo Scientific Alfa Aesar Supplier No. 036189.09
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Zinc arsenide is often used as a component in the manufacture of semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12044-55-2
Molecular Formula As2Zn
Molecular Weight (g/mol) 215.223
MDL Number MFCD00151429
InChI Key KXUSEUKXCIKTBF-UHFFFAOYSA-N
Synonym zinc arsenide znas2, 1,2-diarsa-3-zincacycloprop-1-ene
PubChem CID 102601572
IUPAC Name $l^{2}-arsanylidenearsenic;zinc
SMILES [Zn].[As]=[As]

Specifications

Physical Form Lump
Quantity 10 g
UN Number UN1557
Solubility Information It is insoluble in water.
Formula Weight 215.21
Percent Purity 99.999%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc arsenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.