accessibility menu, dialog, popup

UserName

CAS RN 120512-59-6

CH3FH3COOH

CAS RN 120512-59-6

IUPAC Name: 4-fluoro-3,5-dimethylbenzoic acid
Synonyms: 4-fluoro-3,5-dimethylbenzoic acid
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: MIKQQYBBVDHPST-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results