accessibility menu, dialog, popup

UserName

CAS RN 12061-16-4

IUPAC Name: dierbium(3+) trioxidandiide
Synonyms: erbium(III) oxide
Molecular Weight (g/mol): 382.52
Molecular Formula: Er2O3
InChi Key: ZXGIFJXRQHZCGJ-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Er+3].[Er+3]
Physical Form 
  • (9)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (13)
  • (1)
  • (9)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)