accessibility menu, dialog, popup

UserName

CAS RN 12107-76-5

H3CH2NCH3FBFFF

CAS RN 12107-76-5

IUPAC Name: dibutylazanium; tetrafluoroboranuide
Synonyms: dibutylazanium tetrafluoroborate
Molecular Weight (g/mol): 217.06
Molecular Formula: C8H20BF4N
InChi Key: CAZKFGXFAVXKIF-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.CCCC[NH2+]CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results