missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Di-n-butylammonium tetrafluoroborate, 98%

Product Code. 11489726
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489726 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11489726 Supplier Alfa Aesar™ Supplier No. B20251.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 12107-76-5
Molecular Formula C8H20BF4N
Molecular Weight (g/mol) 217.06
MDL Number MFCD00034899
InChI Key CAZKFGXFAVXKIF-UHFFFAOYSA-O
Synonym di-n-butylammonium tetrafluoroborate, dibutylazanium tetrafluoroborate, dibutylamine tetrafluoroborate, dibutylammonium tetrafluoroborate 1-, dibutylammonium tetrafluoroborate, dibutylammonium, tetrafluoroborate, dibutylamine, tetrafluoroborate 1-, 1-butanamine, n-butyl-, tetrafluoroborate 1-, dibutylamine tetrafluoborate, c8h20n.bf4
PubChem CID 25510
IUPAC Name dibutylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCC[NH2+]CCCC

Specifications

Linear Formula (C4H9)2NH2BF4
Quantity 25 g
Formula Weight 217.06
Percent Purity 98%
Chemical Name or Material Di-n-butylammonium tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.