CAS RN 1211520-62-5
IUPAC Name:
3-(pentafluoro-λ⁶-sulfanyl)benzene-1-sulfonyl chloride
Synonyms:
3-(pentafluoro-λ⁶-sulfanyl)benzenesulfonyl chloride
Molecular Weight (g/mol):
302.66
Molecular Formula:
C6H4ClF5O2S2
InChi Key:
NNEKBNJEZZMVHF-UHFFFAOYSA-N
SMILES:
FS(F)(F)(F)(F)C1=CC=CC(=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
3-(Pentafluorothio)benzenesulfonyl chloride, 97%, Thermo Scientific™
CAS: 1211520-62-5 Molecular Formula: C6H4ClF5O2S2 Molecular Weight (g/mol): 302.658 MDL Number: MFCD16652436 InChI Key: NNEKBNJEZZMVHF-UHFFFAOYSA-N Synonym: 3-pentafluorosulfur benzenesulfonyl chloride,3-pentafluorothio benzenesulfonyl chloride,3-pentafluoro-??-sulfanyl benzenesulfonyl chloride,3-pentafluoro-??-sulfanyl benzene-1-sulfonyl chloride,3-pentafluoro-lambda-sulfanyl benzene-1-sulfonyl chloride PubChem CID: 66523442 IUPAC Name: 3-(pentafluoro-$l^{6}-sulfanyl)benzenesulfonyl chloride SMILES: C1=CC(=CC(=C1)S(F)(F)(F)(F)F)S(=O)(=O)Cl