missing translation for 'onlineSavingsMsg'
Learn More

3-(Pentafluorothio)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15426667
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426667 1 g 1g
15498645 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426667 Supplier Thermo Scientific Alfa Aesar Supplier No. H33799.03
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1211520-62-5
Molecular Formula C6H4ClF5O2S2
Molecular Weight (g/mol) 302.658
MDL Number MFCD16652436
InChI Key NNEKBNJEZZMVHF-UHFFFAOYSA-N
Synonym 3-pentafluorosulfur benzenesulfonyl chloride, 3-pentafluorothio benzenesulfonyl chloride, 3-pentafluoro-??-sulfanyl benzenesulfonyl chloride, 3-pentafluoro-??-sulfanyl benzene-1-sulfonyl chloride, 3-pentafluoro-lambda-sulfanyl benzene-1-sulfonyl chloride
PubChem CID 66523442
IUPAC Name 3-(pentafluoro-$l^{6}-sulfanyl)benzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(F)(F)(F)(F)F)S(=O)(=O)Cl

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 302.66
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.