accessibility menu, dialog, popup

UserName

CH3NHNN

CAS RN 121552-61-2

IUPAC Name: 4-cyclopropyl-6-methyl-N-phenylpyrimidin-2-amine
Synonyms: cyprodinil cga 219417 (cyprodinil)
Molecular Weight (g/mol): 225.3
Molecular Formula: C14H15N3
InChi Key: HAORKNGNJCEJBX-UHFFFAOYSA-N
SMILES: CC1=CC(=NC(NC2=CC=CC=C2)=N1)C1CC1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (6)
Concentration 
  • (3)
  • (3)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
Industry Type 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)