CAS RN 1215621-31-0
CAS RN 1215621-31-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sumatriptan-d6 Succinate, TRC
CAS: 1215621-31-0 Molecular Formula: C14 2H6 H15 N3 O2 S . C4 H6 O4 Molecular Weight (g/mol): 419.53 Synonym: 1-[3-[2-(Bis(trideuteriomethyl)amino)ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide butanedioic acid,3-[2-[Di(methyl-d3)amino]ethyl]-N-methyl-1H-indole-5-methanesulfonamide 1,4-butanedioate (1:1),1-[3-[2-[Di(methyl-d3)amino]ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide succinate,Sumatriptan-d6 succinate,Sumatriptan-D6 (N,N-dimethylamino-D6) Succinate IUPAC Name: 1-[3-[2-[bis(trideuteriomethyl)amino]ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide;butanedioic acid SMILES: [2H]C([2H])([2H])N(CCc1c[nH]c2ccc(CS(=O)(=O)NC)cc12)C([2H])([2H])[2H].OC(=O)CCC(=O)O