missing translation for 'onlineSavingsMsg'
Learn More

Sumatriptan-d6 Succinate, TRC

Product Code. 30426968
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426968 1 mg 1mg
30411152 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30426968 Supplier Toronto Research Chemicals Supplier No. TRCS8100021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215621-31-0
Molecular Formula C14 2H6 H15 N3 O2 S . C4 H6 O4
Molecular Weight (g/mol) 419.53
Synonym 1-[3-[2-(Bis(trideuteriomethyl)amino)ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide butanedioic acid, 3-[2-[Di(methyl-d3)amino]ethyl]-N-methyl-1H-indole-5-methanesulfonamide 1,4-butanedioate (1:1), 1-[3-[2-[Di(methyl-d3)amino]ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide succinate, Sumatriptan-d6 succinate, Sumatriptan-D6 (N,N-dimethylamino-D6) Succinate
IUPAC Name 1-[3-[2-[bis(trideuteriomethyl)amino]ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide;butanedioic acid
SMILES [2H]C([2H])([2H])N(CCc1c[nH]c2ccc(CS(=O)(=O)NC)cc12)C([2H])([2H])[2H].OC(=O)CCC(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H21N3O2S.C4H6O4/c1-15-20(18,19)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2)3;5-3(6)1-2-4(7)8/h4-5,8-9,15-16H,6-7,10H2,1-3H3;1-2H2,(H,5,6)(H,7,8)/i2D3,3D3;
Formula Weight 419.1997
Chemical Name or Material Sumatriptan-d6 Succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.